Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™

Catalog Number AC451370010
Click to view available options
Quantity:
1 g
5 g
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
Boiling Point 120°C
Flash Point 28°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.1
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit