Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
Boiling Point 120°C
Flash Point 28°C
Assay Percent Range 96% min. (GC)
Quantity 5g
Formula Weight 164.1
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene

GHS Signal Word: Warning

RUO – Research Use Only