Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Diisopropylaminoethyl chloride hydrochloride, 98%

Click to view available options
Quantity:
100g
500g
2500g
4261-68-1
C8H19Cl2N
200.147
MFCD00012496
IUSXYVRFJVAVOB-UHFFFAOYSA-N
2-diisopropylaminoethyl chloride hydrochloride,2-diisopropylamino ethyl chloride hydrochloride,unii-6rmr38fceq,2-chloroethyldiisopropylamine hydrochloride,6rmr38fceq,2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride,2-chloroethyldiisopropylammonium chloride,2-chloroethyl diisopropylamine hydrochloride,n-chloroethyl diisopropylamine hydrochloride,beta-chloroethyl diisopropylamine hydrochloride
77942
N-(2-chloroethyl)-N-propan-2-ylpropan-2-amine;hydrochloride
CC(C)N(CCCl)C(C)C.Cl
4261-68-1
C8H19Cl2N
200.147
MFCD00012496
IUSXYVRFJVAVOB-UHFFFAOYSA-N
2-diisopropylaminoethyl chloride hydrochloride,2-diisopropylamino ethyl chloride hydrochloride,unii-6rmr38fceq,2-chloroethyldiisopropylamine hydrochloride,6rmr38fceq,2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride,2-chloroethyldiisopropylammonium chloride,2-chloroethyl diisopropylamine hydrochloride,n-chloroethyl diisopropylamine hydrochloride,beta-chloroethyl diisopropylamine hydrochloride
77942
N-(2-chloroethyl)-N-propan-2-ylpropan-2-amine;hydrochloride
CC(C)N(CCCl)C(C)C.Cl
Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
Melting Point 131°C to 134°C
Quantity 500g
UN Number UN2811
Beilstein 3551603
Formula Weight 200.15
Percent Purity 98%
Chemical Name or Material 2-Diisopropylaminoethyl chloride hydrochloride

GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : TX1521900

TSCA : TSCA