Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)ethyl methacrylate, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
2867-47-2
C8H15NO2
157.21
MFCD00008589
JKNCOURZONDCGV-UHFFFAOYSA-N
2-dimethylamino ethyl methacrylate,dimethylaminoethyl methacrylate,ageflex fm-1,dmaema,usaf rh-3,2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester,2-n,n-dimethylamino ethyl methacrylate,n,n-dimethylaminoethyl methacrylate,2-dimethylaminoethyl methacrylate,n,n-dimethylethanolamine methacrylate
17869
2-(dimethylamino)ethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCN(C)C
2867-47-2
C8H15NO2
157.21
MFCD00008589
JKNCOURZONDCGV-UHFFFAOYSA-N
2-dimethylamino ethyl methacrylate,dimethylaminoethyl methacrylate,ageflex fm-1,dmaema,usaf rh-3,2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester,2-n,n-dimethylamino ethyl methacrylate,n,n-dimethylaminoethyl methacrylate,2-dimethylaminoethyl methacrylate,n,n-dimethylethanolamine methacrylate
17869
2-(dimethylamino)ethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCN(C)C
Viscosity 1.32 mPa.s (20°C)
Specific Gravity 0.933
Flash Point 65°C
Stabilizer 750 to 2000ppm MEHQ
Water 0.1% max. (K.F.)
Infrared Spectrum Authentic
Density 0.9330g/mL
Refractive Index 1.4390 to 1.4410
Color Colorless to Yellow
Packaging Plastic drum
Melting Point -50.0°C
Quantity 5kg
Boiling Point 182.0°C to 192.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)2
Beilstein 04, III, 649
Solubility Information Solubility in water: soluble.
Formula Weight 157.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-(Dimethylamino)ethyl methacrylate, Stabilized
Show More Show Less

GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Warning

EINECSNumber : 220-688-8