Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoethanethiol Hydrochloride 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
13242-44-9
C4H11NS·HCl
141.67
MFCD00012633
NRVFDGZJTPCULU-UHFFFAOYSA-N
2-dimethylamino ethanethiol hydrochloride,captamine hydrochloride,meda,captamine hcl,n,n-dimethylcysteamine hydrochloride,dimethylaminoethanethiol hydrochloride,2-dimethylaminoethanethiol hydrochloride,n-dimethylcysteamine hydrochloride,2-mercaptoethyl dimethylammonium chloride,unii-m43ax41u87
25798
2-(dimethylamino)ethanethiol;hydrochloride
CN(C)CCS.Cl
13242-44-9
C4H11NS·HCl
141.67
MFCD00012633
NRVFDGZJTPCULU-UHFFFAOYSA-N
2-dimethylamino ethanethiol hydrochloride,captamine hydrochloride,meda,captamine hcl,n,n-dimethylcysteamine hydrochloride,dimethylaminoethanethiol hydrochloride,2-dimethylaminoethanethiol hydrochloride,n-dimethylcysteamine hydrochloride,2-mercaptoethyl dimethylammonium chloride,unii-m43ax41u87
25798
2-(dimethylamino)ethanethiol;hydrochloride
CN(C)CCS.Cl
Melting Point 154°C to 159°C
Quantity 5g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (CH3)2NCH2CH2SH·HCl
Solubility Information Solubility in water: soluble in water
Formula Weight 141.67
Chemical Name or Material 2-Dimethylaminoethanethiol hydrochloride, 95%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 236-221-6

RTECSNumber : KJ1051000

TSCA : TSCA