Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoethyl Chloride Hydrochloride 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
4584-46-7,107-99-3
C4H10ClN·HCl
144.05
MFCD00012516
LQLJZSJKRYTKTP-UHFFFAOYSA-N
2-dimethylaminoethyl chloride hydrochloride,2-chloro-n,n-dimethylethanamine hydrochloride,2-dimethylamino ethyl chloride hydrochloride,2-chloroethyl dimethylamine hydrochloride,2-chloro-n,n-dimethylethylamine hydrochloride,dimethylaminoethyl chloride hydrochloride,2-chloroethyldimethylamine hydrochloride,1-chloro-2-dimethylaminoethane hydrochloride,2-dimethylamino ethylchloride hydrochloride,n-2-chloroethyl dimethylamine hydrochloride
11636816
CHEBI:78153
2-chloro-N,N-dimethylethanamine;hydrochloride
CN(C)CCCl.Cl
4584-46-7,107-99-3
C4H10ClN·HCl
144.05
MFCD00012516
LQLJZSJKRYTKTP-UHFFFAOYSA-N
2-dimethylaminoethyl chloride hydrochloride,2-chloro-n,n-dimethylethanamine hydrochloride,2-dimethylamino ethyl chloride hydrochloride,2-chloroethyl dimethylamine hydrochloride,2-chloro-n,n-dimethylethylamine hydrochloride,dimethylaminoethyl chloride hydrochloride,2-chloroethyldimethylamine hydrochloride,1-chloro-2-dimethylaminoethane hydrochloride,2-dimethylamino ethylchloride hydrochloride,n-2-chloroethyl dimethylamine hydrochloride
11636816
CHEBI:78153
2-chloro-N,N-dimethylethanamine;hydrochloride
CN(C)CCCl.Cl
Alkylation reagent
Melting Point 203.0°C to 210.0°C
Packaging Plastic bottle
Water 1% Max. (K.F.)
Quantity 500g
Color Brown-Yellow to White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CH2Cl·HCl
Beilstein 04, 133
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: soluble in alcohol (20°C)
Formula Weight 144.05
Physical Form Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material 2-Dimethylaminoethyl chloride hydrochloride, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed. Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

Warning

EINECSNumber : 224-970-1

RTECSNumber : KQ9020000

TSCA : TSCA