Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
21331-80-6
C22H25BrNP
414.33
MFCD00011794
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
2724136
2-(dimethylamino)ethyl-triphenylphosphanium;bromide
[Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
21331-80-6
C22H25BrNP
414.33
MFCD00011794
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
2724136
2-(dimethylamino)ethyl-triphenylphosphanium;bromide
[Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
for C-C Bond Formation (Olefination)
Quantity 10g
Color Brown-Yellow to White
Infrared Spectrum Authentic
Melting Point 201.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)2NCH2CH2P(C6H5)3Br
Fieser 08,184; 09,291; 13,174; 15,198; 16,205
Formula Weight 414.32
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%
Show More Show Less

EINECSNumber : 244-335-2

TSCA : TSCA