Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoisopropyl chloride hydrochloride, 98.5%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
4584-49-0
C5H12ClN·HCl
158.07
MFCD00012534
OCWGRWAYARCRTQ-UHFFFAOYSA-N
2-dimethylaminoisopropyl chloride hydrochloride,2-chloro-1-dimethylamino propane hydrochloride,2-chloro-n,n-dimethylpropan-1-amine hydrochloride,n,n-dimethyl-2-chloropropylamine hydrochloride,2-chloropropyldimethylammonium chloride,2-dimethylamino isopropyl chloride hydrochloride,2-chloropropyldimethylamine hydrochloride,2-chloropropyl-dimethylamine hydrochloride,1-methyl-2-dimethylaminoethyl chloride hydrochloride,2-chloropropyl dimethylamine hydrochloride
94294
2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
CC(CN(C)C)Cl.Cl
4584-49-0
C5H12ClN·HCl
158.07
MFCD00012534
OCWGRWAYARCRTQ-UHFFFAOYSA-N
2-dimethylaminoisopropyl chloride hydrochloride,2-chloro-1-dimethylamino propane hydrochloride,2-chloro-n,n-dimethylpropan-1-amine hydrochloride,n,n-dimethyl-2-chloropropylamine hydrochloride,2-chloropropyldimethylammonium chloride,2-dimethylamino isopropyl chloride hydrochloride,2-chloropropyldimethylamine hydrochloride,2-chloropropyl-dimethylamine hydrochloride,1-methyl-2-dimethylaminoethyl chloride hydrochloride,2-chloropropyl dimethylamine hydrochloride
94294
2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
CC(CN(C)C)Cl.Cl
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
Loss on Drying 1% max.
Color White to Cream
Melting Point 191.0°C to 195.0°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.5%
Linear Formula CH3CH(Cl)CH2N(CH3)2·HCl
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: soluble in methanol, insoluble in benzene and toluene
Formula Weight 158.07
Physical Form Crystalline Powder
Percent Purity 98.5%
Chemical Name or Material 2-Dimethylaminoisopropyl chloride hydrochloride, 98.5%
Show More Show Less

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

HYGROSCOPIC

EINECSNumber : 224-971-7

RTECSNumber : TY1225000

TSCA : TSCA