Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
42036-65-7
C9H17NO·HCl
191.7
MFCD00102580
CLVHTSWMNNSUSH-UHFFFAOYSA-N
2-dimethylamino methyl cyclohexanone hydrochloride,2-dimethylaminomethyl-1-cyclohexanone hydrochloride,dimethyl 2-oxocyclohexyl methyl ammonium chloride,2-dimethylamino methyl cyclohexan-1-one hydrochloride,2-dimethylaminomethyl-cyclohexanone,2-dimethylamino methyl cyclohexan-1-one, chloride,tramadol related compound b,2-dimethylaminomethyl-cyclohexanone hydrochloride,tramadol related compound b usp
359482
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
CN(C)CC1CCCCC1=O.Cl
42036-65-7
C9H17NO·HCl
191.7
MFCD00102580
CLVHTSWMNNSUSH-UHFFFAOYSA-N
2-dimethylamino methyl cyclohexanone hydrochloride,2-dimethylaminomethyl-1-cyclohexanone hydrochloride,dimethyl 2-oxocyclohexyl methyl ammonium chloride,2-dimethylamino methyl cyclohexan-1-one hydrochloride,2-dimethylaminomethyl-cyclohexanone,2-dimethylamino methyl cyclohexan-1-one, chloride,tramadol related compound b,2-dimethylaminomethyl-cyclohexanone hydrochloride,tramadol related compound b usp
359482
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
CN(C)CC1CCCCC1=O.Cl
Melting Point 147°C to 151°C
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol
Formula Weight 191.7
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%
Show More Show Less