Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diphenylphosphino-2'-(n,n-dimethylamino)biphenyl, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
240417-00-9
C26H24NP
381.459
MFCD03094578
JGFXUYLYPITYGR-UHFFFAOYSA-N
2-diphenylphosphino-2'-n,n-dimethylamino biphenyl,phdave-phos,phdavephos,2-dimethylamino-2'-diphenylphosphino biphenyl,2-2-diphenylphosphanylphenyl-n,n-dimethyl-aniline,2-diphenylphosphino-2;-dimethylaminobiphenyl,1,1'-biphenyl-2-amine, 2'-diphenylphosphino-n,n-dimethyl,2'-diphenylphosphanyl-n,n-dimethyl-1,1'-biphenyl-2-amine
2734939
2-(2-diphenylphosphanylphenyl)-N,N-dimethylaniline
CN(C)C1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
240417-00-9
C26H24NP
381.459
MFCD03094578
JGFXUYLYPITYGR-UHFFFAOYSA-N
2-diphenylphosphino-2'-n,n-dimethylamino biphenyl,phdave-phos,phdavephos,2-dimethylamino-2'-diphenylphosphino biphenyl,2-2-diphenylphosphanylphenyl-n,n-dimethyl-aniline,2-diphenylphosphino-2;-dimethylaminobiphenyl,1,1'-biphenyl-2-amine, 2'-diphenylphosphino-n,n-dimethyl,2'-diphenylphosphanyl-n,n-dimethyl-1,1'-biphenyl-2-amine
2734939
2-(2-diphenylphosphanylphenyl)-N,N-dimethylaniline
CN(C)C1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
Quantity 1g
Melting Point 122°C to 127°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 381.44
Physical Form Powder, Crystals, Crystalline Powder and/or Chunks
Percent Purity ≥96.0%
Chemical Name or Material 2-Diphenylphosphino-2';-(N, N-dimethylamino)biphenyl
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.