Alfa Aesar™ 2-(Diphenylphosphino)benzaldehyde oxime, 95%

Manufacturer: Alfa Aesar™ H5190303

Quantity 1g

 View more versions of this product

Catalog No. AAH5190303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 153358-05-5
    Molecular Formula C19H16NOP
    Formula Weight 305.32
    MDL Number MFCD16251585
    Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde oxime

    Specifications

    Formula Weight 305.32
    Molecular Formula C19H16NOP
    MDL Number MFCD16251585
    CAS 153358-05-5
    Health Hazard 1 Warning
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde oxime
    Assay 95%

    Description

    Applications
    It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

    Solubility
    Insoluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit