Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Diphenylphosphino)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
17261-28-8
C19H15O2P
306.301
MFCD00674024
UYRPRYSDOVYCOU-UHFFFAOYSA-N
2-diphenylphosphino benzoic acid,2-diphenylphosphinobenzoic acid,benzoic acid, 2-diphenylphosphino,o-diphenylphosphinobenozic acid,2-carboxyphenyl diphenylphosphine,2-diphenylphosphanyl benzoic acid,benzoic acid, diphenylphosphino,o-diphenylphosphino benzoic acid,dppbac,acmc-1bt7u
87021
2-diphenylphosphanylbenzoic acid
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3C(=O)O
17261-28-8
C19H15O2P
306.301
MFCD00674024
UYRPRYSDOVYCOU-UHFFFAOYSA-N
2-diphenylphosphino benzoic acid,2-diphenylphosphinobenzoic acid,benzoic acid, 2-diphenylphosphino,o-diphenylphosphinobenozic acid,2-carboxyphenyl diphenylphosphine,2-diphenylphosphanyl benzoic acid,benzoic acid, diphenylphosphino,o-diphenylphosphino benzoic acid,dppbac,acmc-1bt7u
87021
2-diphenylphosphanylbenzoic acid
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3C(=O)O
Quantity 25g
Melting Point 174°C to 181°C
Formula Weight 306.31
Percent Purity 97%
Chemical Name or Material 2-(Diphenylphosphino)benzoic acid

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG9287500

TSCA : TSCA