Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dodecanone, ≥98.5% (GC), MilliporeSigma™ Supelco™

Manufacturer:  MilliporeSigma Supelco 034115ML

 View more versions of this product

Catalog No. 11-101-8367


Add to cart

Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

6175-49-1
MFCD00015064
184.32
Analytical Standard
17°C to 20°C
5 mL
Analytical Standard
CH3(CH2)9 COCH3
Decyl methyl ketone
≥98.5% (GC)
0.828 g/mL (at 20°C)
247°C
108°C
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit