Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dodecanone, ≥98.5% (GC), MilliporeSigma™ Supelco™

$74.20 - $253.00

Specifications

CAS 6175-49-1
Percent Purity ≥98.5% (GC)
Density 0.828 g/mL (at 20°C)
Grade Analytical Standard
Linear Formula CH3(CH2)9 COCH3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9574
View Documents
MilliporeSigma Supelco
034111ML
1 mL
Each for $74.20
Add to cart
 
11-101-8367
View Documents
MilliporeSigma Supelco
034115ML
5 mL
Each for $253.00
Add to cart
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

6175-49-1
0.828 g/mL (at 20°C)
CH3(CH2)9 COCH3
≥98.5% (GC)
Analytical Standard
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit