Alfa Aesar™ 2-Ethoxy-1-naphthaldehyde, 98%

Manufacturer: Alfa Aesar™ B2530606

Quantity 5g

 View more versions of this product

Catalog No. AAB2530606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19523-57-0
    Molecular Formula C13H12O2
    Formula Weight 200.24
    MDL Number MFCD00046427
    Beilstein 1073578
    Synonym 2-Ethoxynaphthalene-1-carboxaldehyde
    Chemical Name or Material 2-Ethoxy-1-naphthaldehyde

    Specifications

    Formula Weight 200.24
    Molecular Formula C13H12O2
    MDL Number MFCD00046427
    CAS 19523-57-0
    Beilstein 1073578
    Synonym 2-Ethoxynaphthalene-1-carboxaldehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    Melting Point 111° to 113°C
    Quantity 5g
    Chemical Name or Material 2-Ethoxy-1-naphthaldehyde
    Assay 98%

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit