Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethoxy-1-naphthoyl chloride, 97%

Click to view available options
Quantity:
1g
5g
25g
55150-29-3
C13H11ClO2
234.679
MFCD00046426
ZRVFFIXOCFUHDA-UHFFFAOYSA-N
2-ethoxy-1-naphthoyl chloride,1-naphthalenecarbonyl chloride, 2-ethoxy,1-naphthalenecarbonylchloride, 2-ethoxy,acmc-209ll8,2-ethoxy naphthoyl chloride,2-ethoxy-1-naphthoylchloride,2-ethoxy-1-naphthalenecarbonyl chloride,2-ethoxy-1-naphthalenecarboxyl chloride,2-ethoxy-naphthalene-1-carbonyl chloride
108679
2-ethoxynaphthalene-1-carbonyl chloride
CCOC1=C(C2=CC=CC=C2C=C1)C(=O)Cl
55150-29-3
C13H11ClO2
234.679
MFCD00046426
ZRVFFIXOCFUHDA-UHFFFAOYSA-N
2-ethoxy-1-naphthoyl chloride,1-naphthalenecarbonyl chloride, 2-ethoxy,1-naphthalenecarbonylchloride, 2-ethoxy,acmc-209ll8,2-ethoxy naphthoyl chloride,2-ethoxy-1-naphthoylchloride,2-ethoxy-1-naphthalenecarbonyl chloride,2-ethoxy-1-naphthalenecarboxyl chloride,2-ethoxy-naphthalene-1-carbonyl chloride
108679
2-ethoxynaphthalene-1-carbonyl chloride
CCOC1=C(C2=CC=CC=C2C=C1)C(=O)Cl
Melting Point 76°C to 80°C
Quantity 25g
UN Number UN3261
Beilstein 2844104
Formula Weight 234.68
Percent Purity 97%
Chemical Name or Material 2-Ethoxy-1-naphthoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA