Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-5-pyridineboronic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
612845-44-0
C7H10BNO3
166.97
UONCERAQKBPLML-UHFFFAOYSA-N
6-ethoxypyridin-3-yl boronic acid,2-ethoxy-5-pyridineboronic acid,2-ethoxypyridine-5-boronic acid,6-ethoxypyridine-3-boronic acid,6-ethoxy-3-pyridinylboronic acid,2-ethoxy-5-pyridylboronic acid,3-borono-6-ethoxypyridine,6-ethoxy-3-pyridylboronic acid,boronic acid, 6-ethoxy-3-pyridinyl,6-ethoxy-3-pyridyl boronic acid
12012237
(6-ethoxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OCC)(O)O
612845-44-0
C7H10BNO3
166.97
UONCERAQKBPLML-UHFFFAOYSA-N
6-ethoxypyridin-3-yl boronic acid,2-ethoxy-5-pyridineboronic acid,2-ethoxypyridine-5-boronic acid,6-ethoxypyridine-3-boronic acid,6-ethoxy-3-pyridinylboronic acid,2-ethoxy-5-pyridylboronic acid,3-borono-6-ethoxypyridine,6-ethoxy-3-pyridylboronic acid,boronic acid, 6-ethoxy-3-pyridinyl,6-ethoxy-3-pyridyl boronic acid
12012237
(6-ethoxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OCC)(O)O
Packaging Glass Bottle
Melting Point 167.0°C to 168.0°C
Infrared Spectrum Authentic
NMR Consistent with structure (1H−NMR)
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 166.97
Percent Purity 97%
Chemical Name or Material 2-Ethoxy-5-pyridineboronic acid

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning