Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide

Manufacturer:  Alfa Aesar™ H3444322

Quantity 100g

 View more versions of this product

Catalog No. AAH3444322


  Check Availability
Add to cart

Specifications

Description

Applications
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

Solubility
Insoluble in water.

Notes
Reacts with strong oxidizing agents. Store at room temperature.
Description & Specifications

Chemical Identifiers

14077-58-8
CH3CH2OCH2COCl
MFCD11170566
2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide
C4H7ClO2
122.55
UN2920

Specifications

14077-58-8
CH3CH2OCH2COCl
MFCD11170566
100g
97%
C4H7ClO2
122.55
UN2920
2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H226-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit