Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethoxybenzamide, 97%

Click to view available options
Quantity:
100g
500g
938-73-8
C9H11NO2
165.192
MFCD00007977
SBNKFTQSBPKMBZ-UHFFFAOYSA-N
ethenzamide,benzamide, 2-ethoxy,o-ethoxybenzamide,ethbenzamide,etenzamide,ethenzamid,etamide,ethosalicyl,etosalicil,etosalicyl
3282
2-ethoxybenzamide
CCOC1=CC=CC=C1C(=O)N
938-73-8
C9H11NO2
165.192
MFCD00007977
SBNKFTQSBPKMBZ-UHFFFAOYSA-N
ethenzamide,benzamide, 2-ethoxy,o-ethoxybenzamide,ethbenzamide,etenzamide,ethenzamid,etamide,ethosalicyl,etosalicil,etosalicyl
3282
2-ethoxybenzamide
CCOC1=CC=CC=C1C(=O)N
Melting Point 131°C to 134°C
Quantity 100g
Merck Index 14,3731
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 2-Ethoxybenzamide

RTECSNumber : CU4381000

TSCA : TSCA