Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzamide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
Plastic bottle
938-73-8
C9H11NO2
165.19
MFCD00007977
SBNKFTQSBPKMBZ-UHFFFAOYSA-N
ethenzamide,benzamide, 2-ethoxy,o-ethoxybenzamide,ethbenzamide,etenzamide,ethenzamid,etamide,ethosalicyl,etosalicil,etosalicyl
3282
2-ethoxybenzamide
CCOC1=CC=CC=C1C(=O)N
938-73-8
C9H11NO2
165.19
MFCD00007977
SBNKFTQSBPKMBZ-UHFFFAOYSA-N
ethenzamide,benzamide, 2-ethoxy,o-ethoxybenzamide,ethbenzamide,etenzamide,ethenzamid,etamide,ethosalicyl,etosalicil,etosalicyl
3282
2-ethoxybenzamide
CCOC1=CC=CC=C1C(=O)N
Color White
Quantity 5g
Melting Point 129.0°C to 134.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5OC6H4CONH2
Beilstein 10, 93
Merck Index 15, 3786
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 165.19
Physical Form Granular Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Ethoxybenzamide, 97%
Show More Show Less

EINECSNumber : 213-346-4

RTECSNumber : CV4900000

TSCA : TSCA