Alfa Aesar™ 2-Ethoxybenzoic acid, 98%

Manufacturer: Alfa Aesar™ 1546818

Quantity 50g

 View more versions of this product

Catalog No. AA1546818

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 134-11-2
    Formula Weight 166.18
    MDL Number MFCD00002438
    Chemical Name or Material 2-Ethoxybenzoic acid

    Specifications

    Formula Weight 166.18
    CAS 134-11-2
    Refractive Index 1.5400
    Melting Point 19°C
    Quantity 50g
    Flash Point 113° (235°F)
    Density 1.105g/mL
    Boiling Point 174° to 176°C (15mmHg)
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00002438
    Chemical Name or Material 2-Ethoxybenzoic acid
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit