Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid, 95%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
133944-74-8
C7H12O3S2
208.29
MFCD19689494
FBDXTAKUEUFSGH-UHFFFAOYSA-N
2-ethoxythioxomethyl thio-2-methylpropanoic acid,2-ethoxythioxo methyl thio-2-methylpropanoic acid,2-ethoxythioxomethyl thio-2-methylpropanoic acid,propanoic acid, 2-ethoxythioxomethyl thio-2-methyl,2-ethoxymethanethioyl sulfanyl-2-methylpropanoic acid,ksc493s4h,2-ethoythioxo methyl thio-isobutyric acid,alpha-ethoxythiocarbonylmercapto-isobutyric acid,2-ethoxycarbonothioylthio-2-methylpropanoic acid,2-ethoxythioxomethyl thio-2-methylpropanoicacid
53249283
2-ethoxycarbothioylsulfanyl-2-methylpropanoic acid
CCOC(=S)SC(C)(C)C(=O)O
133944-74-8
C7H12O3S2
208.29
MFCD19689494
FBDXTAKUEUFSGH-UHFFFAOYSA-N
2-ethoxythioxomethyl thio-2-methylpropanoic acid,2-ethoxythioxo methyl thio-2-methylpropanoic acid,2-ethoxythioxomethyl thio-2-methylpropanoic acid,propanoic acid, 2-ethoxythioxomethyl thio-2-methyl,2-ethoxymethanethioyl sulfanyl-2-methylpropanoic acid,ksc493s4h,2-ethoythioxo methyl thio-isobutyric acid,alpha-ethoxythiocarbonylmercapto-isobutyric acid,2-ethoxycarbonothioylthio-2-methylpropanoic acid,2-ethoxythioxomethyl thio-2-methylpropanoicacid
53249283
2-ethoxycarbothioylsulfanyl-2-methylpropanoic acid
CCOC(=S)SC(C)(C)C(=O)O
Quantity 5g
Beilstein 1779889
Formula Weight 208.29
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning