Alfa Aesar™ 2-Ethoxycyclohexanone, 98%

Manufacturer: Alfa Aesar™ H2761303

Quantity 1g

 View more versions of this product

Catalog No. AAH2761303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 33371-97-0
    Molecular Formula C8H14O2
    Formula Weight 142.2
    MDL Number MFCD00015377
    UN Number UN1224
    Chemical Name or Material 2-Ethoxycyclohexanone

    Specifications

    Formula Weight 142.2
    Molecular Formula C8H14O2
    MDL Number MFCD00015377
    UN Number UN1224
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Flash Point 54°C (129°F)
    Boiling Point 85°C (15mmHg)
    Quantity 1g
    CAS 33371-97-0
    Chemical Name or Material 2-Ethoxycyclohexanone
    Assay 98%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit