Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethanol, for analysis, Acros Organics

Click to view available options
Quantity:
1L
2.5L
110-80-5
C4H10O2
90.12
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve,oxitol,ethyl cellosolve,ethylene glycol monoethyl ether,ethanol, 2-ethoxy,ethyl glycol,emkanol,hydroxy ether,ethylene glycol ethyl ether,dowanol ee
8076
CHEBI:46788
2-ethoxyethanol
CCOCCO
110-80-5
C4H10O2
90.12
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve,oxitol,ethyl cellosolve,ethylene glycol monoethyl ether,ethanol, 2-ethoxy,ethyl glycol,emkanol,hydroxy ether,ethylene glycol ethyl ether,dowanol ee
8076
CHEBI:46788
2-ethoxyethanol
CCOCCO
Viscosity 2.08 mPa.s (20°C)
Melting Point -70°C
Flash Point 40°C
Quantity 2.5L
Water 0.1% max.
Refractive Index 1.4060 to 1.4100
Infrared Spectrum Authentic
Density 0.9300g/mL
Boiling Point 135.0°C
Residue after Evaporation 0.005% max.
Specific Gravity 0.93
Linear Formula C2H5OCH2CH2OH
Merck Index 15,3803
Solubility Information Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
Formula Weight 90.12
Percent Purity ≥98.5% (GC)
Grade Analysis
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
May damage fertility.
May damage the unborn child.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 203-804-1