Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl Ether, 98+%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Metal drum
112-36-7
C8H18O3
162.23
MFCD00009254
RRQYJINTUHWNHW-UHFFFAOYSA-N
diethylene glycol diethyl ether,2-ethoxyethyl ether,diethyl carbitol,ethyl diglyme,1-ethoxy-2-2-ethoxyethoxy ethane,bis 2-ethoxyethyl ether,diethyldiethylene glycol,3,6,9-trioxaundecane,degdee,ether, bis 2-ethoxyethyl
8179
CHEBI:44664
1-ethoxy-2-(2-ethoxyethoxy)ethane
CCOCCOCCOCC
112-36-7
C8H18O3
162.23
MFCD00009254
RRQYJINTUHWNHW-UHFFFAOYSA-N
diethylene glycol diethyl ether,2-ethoxyethyl ether,diethyl carbitol,ethyl diglyme,1-ethoxy-2-2-ethoxyethoxy ethane,bis 2-ethoxyethyl ether,diethyldiethylene glycol,3,6,9-trioxaundecane,degdee,ether, bis 2-ethoxyethyl
8179
CHEBI:44664
1-ethoxy-2-(2-ethoxyethoxy)ethane
CCOCCOCCOCC
Boiling Point 180°C to 190°C
Melting Point -44.3°C
Flash Point 82°C
Density 0.9000g/mL
Specific Gravity 0.9
Viscosity 1.4 mPa.s (20°C)
Refractive Index 1.4105 to 1.4125
Residue after Evaporation 0.05% max.
Quantity 2.5kg
Infrared Spectrum Authentic
Packaging Metal drum
Color Undesignated
Water 0.2% max. (K.F.)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (C2H5OCH2CH2)2O
Beilstein 01, II, 519
Merck Index 15, 3132
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in alcohol and other organic solvents,soluble in hydrocarbons,soluble in ether
Formula Weight 162.23
Physical Form Liquid
Percent Purity ≥98.0%
Grade Extra Pure
Chemical Name or Material 2-Ethoxyethyl ether
Show More Show Less

GHS H Statement:
May form explosive peroxides.

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.