Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethoxynaphthalene, 99%

Click to view available options
Quantity:
25g
100g
500g
93-18-5
C12H12O
172.227
MFCD00016808
GUMOJENFFHZAFP-UHFFFAOYSA-N
bromelia,naphthalene, 2-ethoxy,neroline,nerolin new,nerolin ii,ethyl 2-naphthyl ether,2-naphthol ethyl ether,bromelia compound,beta-naphthyl ethyl ether,ethyl beta-naphthyl ether
7129
2-ethoxynaphthalene
CCOC1=CC2=CC=CC=C2C=C1
93-18-5
C12H12O
172.227
MFCD00016808
GUMOJENFFHZAFP-UHFFFAOYSA-N
bromelia,naphthalene, 2-ethoxy,neroline,nerolin new,nerolin ii,ethyl 2-naphthyl ether,2-naphthol ethyl ether,bromelia compound,beta-naphthyl ethyl ether,ethyl beta-naphthyl ether
7129
2-ethoxynaphthalene
CCOC1=CC2=CC=CC=C2C=C1
Melting Point 35°C to 37°C
Quantity 500g
Boiling Point 282°C
Flash Point 134°C (273°F)
Beilstein 2042397
Merck Index 14,3752
Formula Weight 172.23
Percent Purity 99%
Chemical Name or Material 2-Ethoxynaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ6900000

TSCA : TSCA