Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxynaphthylidene-4-fluoroacetophenone, 98%, Acros Organics

Click to view available options
Quantity:
5g
-01-1
C21H17FO2
MFCD03788502
-01-1
C21H17FO2
MFCD03788502
Quantity 5g
Color Yellow
Melting Point 132°C to 136°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 320.36
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Ethoxynaphthylidene-4-fluoroacetophenone, 98%