Alfa Aesar™ 2-Ethoxyphenol, 98%

Manufacturer: Alfa Aesar™ B2475522

Quantity 100g

 View more versions of this product

Catalog No. AAB2475522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94-71-3
    Molecular Formula C8H10O2
    Formula Weight 138.17
    MDL Number MFCD00002187
    Synonym Catechol monoethyl ether
    Chemical Name or Material 2-Ethoxyphenol

    Specifications

    Formula Weight 138.17
    Odor Phenol-like
    TSCA TSCA
    Molecular Formula C8H10O2
    MDL Number MFCD00002187
    Synonym Catechol monoethyl ether
    Health Hazard 2 GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Flash Point 86°C (187°F)
    Boiling Point 214° to 216°C
    Melting Point 24° to 27°C
    Quantity 100g
    CAS 94-71-3
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Ethoxyphenol
    Assay 98%

    GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit