Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyphenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
94-71-3
C8H10O2
138.17
MFCD00002187
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
o-ethoxyphenol,phenol, 2-ethoxy,guaethol,guethol,2-ethyloxyphenol,guaiethol,pyrocatechol monoethyl ether,catechol monoethyl ether,phenol, o-ethoxy,unii-878iw8p9pw
66755
2-ethoxyphenol
CCOC1=CC=CC=C1O
94-71-3
C8H10O2
138.17
MFCD00002187
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
o-ethoxyphenol,phenol, 2-ethoxy,guaethol,guethol,2-ethyloxyphenol,guaiethol,pyrocatechol monoethyl ether,catechol monoethyl ether,phenol, o-ethoxy,unii-878iw8p9pw
66755
2-ethoxyphenol
CCOC1=CC=CC=C1O
Specific Gravity 1.09
Packaging Glass bottle
Density 1.0900g/mL
Boiling Point 216.0°C to 217.0°C
Infrared Spectrum Authentic
Refractive Index 1.5280 to 1.5300
Melting Point 20.0°C to 25.0°C
Color Colorless to Yellow
Quantity 5g
Flash Point 91°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C2H5OC6H4OH
Formula Weight 138.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Ethoxyphenol
Show More Show Less

GHS H Statement
May cause an allergic skin reaction. Causes serious eye damage. Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Gently wash with plenty of soap and water. Avoid release to the environment.

Danger