Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxypyridine-3-boronic acid pinacol ester, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
5g
848243-23-2
C13H20BNO3
249.12
GRXNEZGBXVIORK-UHFFFAOYSA-N
2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine,2-ethoxypyridine-3-boronic acid pinacol ester,2-ethoxypyridin-3-ylboronic acid pinacol ester,2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pubchem16583,2-ethoxypyridin-3-yl boronic acid pinacol ester
17750191
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)OCC
848243-23-2
C13H20BNO3
249.12
GRXNEZGBXVIORK-UHFFFAOYSA-N
2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine,2-ethoxypyridine-3-boronic acid pinacol ester,2-ethoxypyridin-3-ylboronic acid pinacol ester,2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pubchem16583,2-ethoxypyridin-3-yl boronic acid pinacol ester
17750191
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)OCC
Infrared Spectrum Authentic
Quantity 250mg
Assay Percent Range 94% min. (GC)
Formula Weight 249.12
Percent Purity 95%
Chemical Name or Material 2-Ethoxypyridine-3-boronic acid pinacol ester

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning