Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1,3-hexanediol, Mixture of Isomers 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
94-96-2
C8H18O2
146.23
MFCD00004578
RWLALWYNXFYRGW-UHFFFAOYSA-N
2-ethyl-1,3-hexanediol,ethohexadiol,1,3-hexanediol, 2-ethyl,octylene glycol,ethyl hexanediol,carbide 6-12,repellent 612,rutgers 612,6-12-insect repellent,diol-kyowa 8
7211
CHEBI:34273
2-ethylhexane-1,3-diol
CCCC(C(CC)CO)O
94-96-2
C8H18O2
146.23
MFCD00004578
RWLALWYNXFYRGW-UHFFFAOYSA-N
2-ethyl-1,3-hexanediol,ethohexadiol,1,3-hexanediol, 2-ethyl,octylene glycol,ethyl hexanediol,carbide 6-12,repellent 612,rutgers 612,6-12-insect repellent,diol-kyowa 8
7211
CHEBI:34273
2-ethylhexane-1,3-diol
CCCC(C(CC)CO)O
Refractive Index 1.4495 to 1.4525
Melting Point -40°C
Boiling Point 243°C
Flash Point 136°C
Viscosity 323 mPa.s (20°C)
Specific Gravity 0.9422
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.2% max.
Acidity 0.05mg KOH/g max.
Quantity 1L
Density 0.9422g/mL
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH2CH2CH(OH)CH(C2H5)CH2OH
Merck Index 15, 3797
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in ethanol,isopropanol,propylene glycol,,castor oil and ether
Formula Weight 146.23
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Ethyl-1, 3-hexanediol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-377-9

RTECSNumber : MO2625000

TSCA : TSCA