Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-1H-indene, 97%

Click to view available options
Quantity:
1g
5g
25g
17059-50-6
C11H12
144.217
MFCD01321239
BSHJHVHMLRKHBZ-UHFFFAOYSA-N
2-ethylindene,2-ethyl-1-h-indene,1h-indene, 2-ethyl,pubchem5396,1h-indene,2-ethyl,acmc-20a5x3
582328
2-ethyl-1H-indene
CCC1=CC2=CC=CC=C2C1
17059-50-6
C11H12
144.217
MFCD01321239
BSHJHVHMLRKHBZ-UHFFFAOYSA-N
2-ethylindene,2-ethyl-1-h-indene,1h-indene, 2-ethyl,pubchem5396,1h-indene,2-ethyl,acmc-20a5x3
582328
2-ethyl-1H-indene
CCC1=CC2=CC=CC=C2C1
Flash Point 87°C (190°F)
Quantity 1g
Boiling Point 94°C to 96°C (10mmHg)
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1H-indene

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning