Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-phenylmalonamide hydrate, 97%, contains max. 25% water, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7206-76-0
C11H14N2O2
206.245
MFCD00149557
JFZHPFOXAAIUMB-UHFFFAOYSA-N
23611
CHEBI:8097
2-ethyl-2-phenylpropanediamide
CCC(C1=CC=CC=C1)(C(=O)N)C(=O)N
7206-76-0
C11H14N2O2
206.245
MFCD00149557
JFZHPFOXAAIUMB-UHFFFAOYSA-N
23611
CHEBI:8097
2-ethyl-2-phenylpropanediamide
CCC(C1=CC=CC=C1)(C(=O)N)C(=O)N
Quantity 1g
Color White to Pink
Packaging Glass bottle
Loss on Drying 25% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C6H5C(C2H5)(CONH2)2·xH2O
Formula Weight 206.24
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Ethyl-2-phenylmalonamide hydrate
Show More Show Less