Alfa Aesar™ 2-Ethyl-2-vinyl-1,3-dioxolane, 98%

Manufacturer: Alfa Aesar™ H3197109

Quantity 10g

 View more versions of this product

Catalog No. AAH3197109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22515-82-8
    Molecular Formula C7H12O2
    Formula Weight 128.17
    MDL Number MFCD13185932
    UN Number UN3271
    Chemical Name or Material 2-Ethyl-2-vinyl-1,3-dioxolane

    Specifications

    Formula Weight 128.17
    Molecular Formula C7H12O2
    MDL Number MFCD13185932
    UN Number UN3271
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 70°C (77mmHg)
    Odor Characteristic
    Quantity 10g
    CAS 22515-82-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    Chemical Name or Material 2-Ethyl-2-vinyl-1,3-dioxolane
    Assay 98%

    Description

    Applications
    It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

    Solubility
    Insoluble in water.

    Notes
    Store in cool dry place. Keep away from oxidizing agents.

    GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit