Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-2-vinyl-1,3-dioxolane, 98%

Manufacturer:  Alfa Aesar™ H3197118

Quantity 50g

 View more versions of this product

Catalog No. AAH3197118


  Check Availability
Add to cart

Specifications

Description

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

22515-82-8
128.17
UN3271
C7H12O2
MFCD13185932
2-Ethyl-2-vinyl-1,3-dioxolane

Specifications

22515-82-8
128.17
UN3271
Characteristic
2-Ethyl-2-vinyl-1,3-dioxolane
C7H12O2
MFCD13185932
70°C (77mmHg)
50g
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit