Alfa Aesar™ 2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers

Manufacturer: Alfa Aesar™ H2738322

Quantity 100g

 View more versions of this product

Catalog No. AAH2738322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13925-07-0
    Molecular Formula C8H12N2
    Formula Weight 136.19
    MDL Number MFCD00047392
    Chemical Name or Material 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers

    Specifications

    Formula Weight 136.19
    TSCA TSCA
    Molecular Formula C8H12N2
    MDL Number MFCD00047392
    CAS 13925-07-0
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P264-P301+P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 182°C
    Quantity 100g
    Flash Point 66°C (151°F)
    Chemical Name or Material 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers
    Assay 99%

    Description

    Applications
    It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.

    Solubility
    Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.

    GHS P Statement
    P210-P280-P264-P301+P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit