Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-3-methoxybenzoic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
57598-51-3
C10H12O3
180.203
MFCD11100772
RXCOEMMGBRUEEG-UHFFFAOYSA-N
2-ethyl-3-methoxybenzoicacid,2-ethyl-3-methoxy-benzoic acid,benzoic acid, 2-ethyl-3-methoxy,pubchem22099,3-methoxy-2-ethylbenzoicacid,ksc495k0f
12247212
2-ethyl-3-methoxybenzoic acid
CCC1=C(C=CC=C1OC)C(=O)O
57598-51-3
C10H12O3
180.203
MFCD11100772
RXCOEMMGBRUEEG-UHFFFAOYSA-N
2-ethyl-3-methoxybenzoicacid,2-ethyl-3-methoxy-benzoic acid,benzoic acid, 2-ethyl-3-methoxy,pubchem22099,3-methoxy-2-ethylbenzoicacid,ksc495k0f
12247212
2-ethyl-3-methoxybenzoic acid
CCC1=C(C=CC=C1OC)C(=O)O
Quantity 1g
Beilstein 2804017
Formula Weight 180.2
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Ethyl-3-methoxybenzoic acid

storageNote1 : -30° to -10°C