Alfa Aesar™ 2-Ethyl-4,5-dimethyl-3-thiazoline, cis + trans, 99%

Manufacturer: Alfa Aesar™ H2696003

Quantity 1g

 View more versions of this product

Catalog No. AAH2696003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76788-46-0
    Molecular Formula C7H13NS
    Formula Weight 143.26
    MDL Number MFCD01680822
    Synonym 4,5-Dimethyl-2-ethyl-3-thiazoline
    Chemical Name or Material 2-Ethyl-4,5-dimethyl-3-thiazoline, Cis + Trans

    Specifications

    Formula Weight 143.26
    Molecular Formula C7H13NS
    MDL Number MFCD01680822
    Synonym 4,5-Dimethyl-2-ethyl-3-thiazoline
    Health Hazard 2 GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P264-P301+P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 65°C (3mmHg)
    Quantity 1g
    Flash Point 65°C (149°F)
    Recommended Storage 2° to 8°C
    RTECS Number XJ6642500
    CAS 76788-46-0
    Health Hazard 1 Warning
    Chemical Name or Material 2-Ethyl-4,5-dimethyl-3-thiazoline, Cis + Trans
    Assay 99%

    GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P280-P264-P301+P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit