Alfa Aesar™ 2-Ethyl-4,5-dimethyloxazole, 98%

Manufacturer: Alfa Aesar™ H2747006

Quantity 5g

 View more versions of this product

Catalog No. AAH2747006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53833-30-0
    Molecular Formula C7H11NO
    Formula Weight 125.18
    MDL Number MFCD00036664
    Chemical Name or Material 2-Ethyl-4,5-dimethyloxazole

    Specifications

    Formula Weight 125.18
    TSCA TSCA
    Molecular Formula C7H11NO
    MDL Number MFCD00036664
    CAS 53833-30-0
    Quantity 5g
    Chemical Name or Material 2-Ethyl-4,5-dimethyloxazole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit