Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-5,5-dimethyl-1,3-dioxane, 99%

Manufacturer:  Alfa Aesar™ H3235114

Quantity 25g

 View more versions of this product

Catalog No. AAH3235114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

768-58-1
144.21
UN1993
C8H16O2
MFCD16622268
2-Ethyl-5,5-dimethyl-1,3-dioxane

Specifications

768-58-1
144.21
UN1993
Characteristic
50°C (122°F)
99%
C8H16O2
MFCD16622268
154°C
25g
2-Ethyl-5,5-dimethyl-1,3-dioxane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit