Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-5-methylthiophene, 99%

Click to view available options
Quantity:
1g
5g
25g
40323-88-4
C7H10S
126.217
MFCD00053128
VOIVNYVBGCJFRW-UHFFFAOYSA-N
thiophene, 2-ethyl-5-methyl,2-ethyl-5-methyl-thiophene,2-methyl-5-ethyl-thiophene,2-methyl-5-ethylthiophene,5-methyl-2-ethylthiophene,acmc-209jd4,thiophene,2-ethyl-5-methyl
520990
2-ethyl-5-methylthiophene
CCC1=CC=C(S1)C
40323-88-4
C7H10S
126.217
MFCD00053128
VOIVNYVBGCJFRW-UHFFFAOYSA-N
thiophene, 2-ethyl-5-methyl,2-ethyl-5-methyl-thiophene,2-methyl-5-ethyl-thiophene,2-methyl-5-ethylthiophene,5-methyl-2-ethylthiophene,acmc-209jd4,thiophene,2-ethyl-5-methyl
520990
2-ethyl-5-methylthiophene
CCC1=CC=C(S1)C
Quantity 1g
UN Number UN1993
Formula Weight 126.22
Percent Purity 99%
Chemical Name or Material 2-Ethyl-5-methylthiophene

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.