Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethyl-6-methylaniline, 98%

Click to view available options
Quantity:
500g
2500g
24549-06-2
C9H13N
135.21
MFCD00007752
JJVKJJNCIILLRP-UHFFFAOYSA-N
6-ethyl-o-toluidine,2-methyl-6-ethylaniline,benzenamine, 2-ethyl-6-methyl,o-toluidine, 6-ethyl,6-ethyl-2-toluidine,2-methyl-6-ethyl aniline,aniline, 2-methyl-6-ethyl,6-ethyl-2-methylaniline,2-ethyl-6-methyl-aniline,2-ethyl-6-methylbenzenamine
32485
2-ethyl-6-methylaniline
CCC1=CC=CC(=C1N)C
24549-06-2
C9H13N
135.21
MFCD00007752
JJVKJJNCIILLRP-UHFFFAOYSA-N
6-ethyl-o-toluidine,2-methyl-6-ethylaniline,benzenamine, 2-ethyl-6-methyl,o-toluidine, 6-ethyl,6-ethyl-2-toluidine,2-methyl-6-ethyl aniline,aniline, 2-methyl-6-ethyl,6-ethyl-2-methylaniline,2-ethyl-6-methyl-aniline,2-ethyl-6-methylbenzenamine
32485
2-ethyl-6-methylaniline
CCC1=CC=CC(=C1N)C
Quantity 500g
Flash Point 89°C (192°F)
Refractive Index 1.552
Sensitivity Air and light sensitive
Density 0.969
Boiling Point 226°C to 228°C
Melting Point -25°C
Beilstein 2079468
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material 2-Ethyl-6-methylaniline
Show More Show Less

GHS H Statement
H301-H311-H227-H332
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.

P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P311-P312-P330-P362-P374-P501c

H227-H302+H312-H331

EINECSNumber : 246-309-6

RTECSNumber : BY5600000

TSCA : Yes

Recommended Storage : Ambient temperatures