Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-6-methyliodobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
175277-95-9
C9H11I
246.091
MFCD00052855
PCSPENCTMKMGFE-UHFFFAOYSA-N
2-ethyl-6-methyliodobenzene,3-ethyl-2-iodotoluene,acmc-1c540,1-methyl-2-iodo-3-ethylbenzene,3-ethyl-2-iodo-1-methylbenzene,benzene, 1-ethyl-3-methyl-2-iodo
2734195
1-ethyl-2-iodo-3-methylbenzene
CCC1=CC=CC(=C1I)C
175277-95-9
C9H11I
246.091
MFCD00052855
PCSPENCTMKMGFE-UHFFFAOYSA-N
2-ethyl-6-methyliodobenzene,3-ethyl-2-iodotoluene,acmc-1c540,1-methyl-2-iodo-3-ethylbenzene,3-ethyl-2-iodo-1-methylbenzene,benzene, 1-ethyl-3-methyl-2-iodo
2734195
1-ethyl-2-iodo-3-methylbenzene
CCC1=CC=CC(=C1I)C
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 1g
Color Undesignated
Packaging Glass Bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H3(I)C2H5
Formula Weight 246.09
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Ethyl-6-methyliodobenzene
Show More Show Less

LIGHT SENSITIVE

TSCA : TSCA