Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone

Click to view available options
Quantity:
5g
25g
100g
922-63-4
C5H8O
84.118
MFCD00010129
GMLDCZYTIPCVMO-UHFFFAOYSA-N
2-ethylacrolein,2-methylenebutanal,2-ethylpropenal,butanal, 2-methylene,ethacrolein,2-ethylacrylaldehyde,2-methylenebutyraldehyde,ethylacrolein,alpha-ethylacrolein,alpha-ethylacrylaldehyde
70203
2-methylidenebutanal
CCC(=C)C=O
922-63-4
C5H8O
84.118
MFCD00010129
GMLDCZYTIPCVMO-UHFFFAOYSA-N
2-ethylacrolein,2-methylenebutanal,2-ethylpropenal,butanal, 2-methylene,ethacrolein,2-ethylacrylaldehyde,2-methylenebutyraldehyde,ethylacrolein,alpha-ethylacrolein,alpha-ethylacrylaldehyde
70203
2-methylidenebutanal
CCC(=C)C=O
Applications
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
Odor Pungent
Quantity 5g
Boiling Point 92°C to 93°C
Flash Point 1°C (34°F)
UN Number UN1992
Beilstein 1098378
Formula Weight 84.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Ethylacrolein, stabilized with 50ppm hydroquinone

GHS H Statement
H225-H330-H341-H315-H319-H335
Highly flammable liquid and vapour.
Fatal if inhaled.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : ES3480000

Recommended Storage : 2° to 8°C