Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylanthraquinone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
84-51-5
C16H12O2
236.27
MFCD00001237
SJEBAWHUJDUKQK-UHFFFAOYSA-N
2-ethylanthraquinone,2-ethyl anthraquinone,2-ethyl-9,10-anthraquinone,anthraquinone, 2-ethyl,9,10-anthracenedione, 2-ethyl,usaf so-1,beta-ethylanthraquinone,unii-59yj81qzkd,59yj81qzkd,2-ethylanthra-9,10-quinone
6772
2-ethylanthracene-9,10-dione
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
84-51-5
C16H12O2
236.27
MFCD00001237
SJEBAWHUJDUKQK-UHFFFAOYSA-N
2-ethylanthraquinone,2-ethyl anthraquinone,2-ethyl-9,10-anthraquinone,anthraquinone, 2-ethyl,9,10-anthracenedione, 2-ethyl,usaf so-1,beta-ethylanthraquinone,unii-59yj81qzkd,59yj81qzkd,2-ethylanthra-9,10-quinone
6772
2-ethylanthracene-9,10-dione
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
Packaging Glass bottle
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Melting Point 108°C to 111°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 07, I, 425
Formula Weight 236.27
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Ethylanthraquinone, 97%
Show More Show Less

EINECSNumber : 201-535-4

RTECSNumber : CB0525000

TSCA : TSCA