Alfa Aesar™ 2-Ethylbutyraldehyde, 98%

Manufacturer: Alfa Aesar™ B23465AK

Quantity 250mL

 View more versions of this product

Catalog No. AAB23465AK

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 97-96-1
    Molecular Formula C6H12O
    Formula Weight 100.16
    MDL Number MFCD00006985
    UN Number UN1178
    Beilstein 1209330
    Chemical Name or Material 2-Ethylbutyraldehyde

    Specifications

    Formula Weight 100.16
    TSCA TSCA
    Boiling Point 116° to 118°C
    Molecular Formula C6H12O
    Quantity 250mL
    Flash Point 8°C (46°F)
    Odor Like Aldehyde
    MDL Number MFCD00006985
    UN Number UN1178
    Beilstein 1209330
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H319-H335
    Highly flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number ES2625000
    CAS 97-96-1
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-ETHYLBUTYRALDEHYDE
    Chemical Name or Material 2-Ethylbutyraldehyde
    Assay 98%

    Description

    Applications
    2-Ethylbutyraldehyde is used in the preparation of aldoximes using aqueous hydroxylamine. It is used as pharmaceutical intermediates.

    Solubility
    Soluble in ether and alcohols. Slightly soluble in water.

    Notes
    Air Sensitive. Store under dry inert gas. Store away from air.

    GHS H Statement
    H225-H319-H335
    Highly flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit