Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbutyric Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
88-09-5
C6H12O2
116.16
MFCD00002670
OXQGTIUCKGYOAA-UHFFFAOYSA-N
2-ethylbutyric acid,diethylacetic acid,butanoic acid, 2-ethyl,acetic acid, diethyl,3-pentanecarboxylic acid,2-ethyl butanoic acid,diethyl acetic acid,butyric acid, 2-ethyl,2-ethyl-butyric acid,alpha-ethylbutyric acid
6915
2-ethylbutanoic acid
CCC(CC)C(=O)O
88-09-5
C6H12O2
116.16
MFCD00002670
OXQGTIUCKGYOAA-UHFFFAOYSA-N
2-ethylbutyric acid,diethylacetic acid,butanoic acid, 2-ethyl,acetic acid, diethyl,3-pentanecarboxylic acid,2-ethyl butanoic acid,diethyl acetic acid,butyric acid, 2-ethyl,2-ethyl-butyric acid,alpha-ethylbutyric acid
6915
2-ethylbutanoic acid
CCC(CC)C(=O)O
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Density 0.9200g/mL
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C2H5)2CHCO2H
Beilstein 02, 333
Merck Index 15, 3121
Formula Weight 116.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Ethylbutyric acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-796-4

RTECSNumber : ET1400000

TSCA : TSCA