Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

2-Ethylhexanoic acid is one of the di(2-ethylhexyl)phthalate (DEHP) metabolites.

$27.50 - $91.00

Specifications

CAS 149-57-5
Type Analytical Standard
Linear Formula CH3(CH2)3 CH(C2H5)CO2 H
MDL Number MFCD00002675
Synonym 2-Ethylcaproic acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3186
View Documents
MilliporeSigma Supelco
025291MLF
1 mL
Each for $27.50
Add to Cart
 
11-101-3187
View Documents
MilliporeSigma Supelco
025295MLF
5 mL
Each for $91.00
Add to Cart
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

149-57-5
CH3(CH2)3 CH(C2H5)CO2 H
2-Ethylcaproic acid
≥99.5% (GC)
699°F
4.98 (vs. air)
228°C
Analytical Standard
MFCD00002675
144.21
Analytical Standard
10 mmHg (115°C)
0.903 g/mL (at 25°C)
114°C - Closed Cup
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : MO7700000

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit