Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexanoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
1L
2.5kg
10kg
Packaging:
Glass Bottle
Plastic drum
149-57-5
C8H16O2
144.21
MFCD00002675
OBETXYAYXDNJHR-UHFFFAOYSA-N
2-ethylcaproic acid,hexanoic acid, 2-ethyl,ethylhexanoic acid,ethylhexoic acid,2-ethylhexoic acid,butylethylacetic acid,2-butylbutanoic acid,3-heptanecarboxylic acid,ethyl hexanoic acid,2-ethyl-hexoic acid
8697
2-ethylhexanoic acid
CCCCC(CC)C(=O)O
149-57-5
C8H16O2
144.21
MFCD00002675
OBETXYAYXDNJHR-UHFFFAOYSA-N
2-ethylcaproic acid,hexanoic acid, 2-ethyl,ethylhexanoic acid,ethylhexoic acid,2-ethylhexoic acid,butylethylacetic acid,2-butylbutanoic acid,3-heptanecarboxylic acid,ethyl hexanoic acid,2-ethyl-hexoic acid
8697
2-ethylhexanoic acid
CCCCC(CC)C(=O)O
Viscosity 7.7 mPa.s (20°C)
Refractive Index 1.4240 to 1.4260
Water 0.1% max. (K.F.)
Melting Point -59.0°C
Specific Gravity 0.906
Packaging Plastic drum
Density 0.9060g/mL
Boiling Point 228.0°C
Quantity 2.5kg
Color Undesignated
Flash Point 114°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)3CH(C2H5)CO2H
Beilstein 02,349
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in ether
Formula Weight 144.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Ethylhexanoic acid
Show More Show Less

GHS H Statement:
Suspected of damaging the unborn child.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning