Alfa Aesar™ 2-Ethylhexyl acetate, 99%

Manufacturer: Alfa Aesar™ B23257AK

Quantity 250mL

 View more versions of this product

Catalog No. AAB23257AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 103-09-3
    Molecular Formula C10H20O2
    Formula Weight 172.27
    MDL Number MFCD00027249
    Beilstein 1758321
    Merck Index 14,6763
    Synonym Acetic acid 2-ethylhexyl ester; ''Octyl'' acetate
    Chemical Name or Material 2-Ethylhexyl acetate

    Specifications

    Formula Weight 172.27
    TSCA TSCA
    Odor Pleasant
    Molecular Formula C10H20O2
    MDL Number MFCD00027249
    CAS 103-09-3
    Merck Index 14,6763
    Synonym Acetic acid 2-ethylhexyl ester; ''Octyl'' acetate
    Boiling Point 199° to 200°C
    Melting Point −45°C
    Quantity 250mL
    Flash Point 76°C (169°F)
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number AH5600000
    Beilstein 1758321
    Health Hazard 1 Warning
    Chemical Name or Material 2-Ethylhexyl acetate
    Assay 99%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit